missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Tyrosine Antibody (PY793), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-34730AF594
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in All Species samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Especificaciones
Phospho-Tyrosine | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Protein phosphorylation is a fundamental event in the regulation of a large number of intracellular processes. Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be auto-phosphorylated kinases or certain cellular protein substrates. Tyrosine-phosphorylated proteins are involved in signal transduction and in the regulation of cell proliferation. Antibody to phosphotyrosine provides an excellent tool for the detection, characterization, and purification of phosphotyrosine containing proteins. This monoclonal antibody shows no cross-reaction with other phosphoamino acids and is superb for multiple applications including staining of formalin/paraffin tissues. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PY793 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine | |
Phosphotyrosine conjugated to BSA | |
0.1 mL | |
Primary | |
Human, All species | |
IgG2b κ |