Learn More
Eurofins DiscoverX PI3 Kinase (p110α/p85α) Mouse Protein
Complex of N-terminal 6His-tagged, recombinant, full-length mouse p110α and untagged, recombinant, full length mouse p85α. Coexpressed by baculovirus in Sf21 insect cells, for use in Kinase Assays.
Marca: Eurofins DiscoverX 14-785M
Detalles adicionales : Peso : 4.70710kg
Descripción
PIP3 plays a key role by recruiting PH domain-containing proteins to the membrane, including AKT1 and PDPK1, activating signaling cascades involved in cell growth, survival, proliferation, motility and morphology. Participates in cellular signaling in response to various growth factors. Involved in the activation of AKT1 upon stimulation by receptor tyrosine kinases ligands such as EGF, insulin, IGF1, VEGFA and PDGF. Involved in signaling via insulin-receptor substrate (IRS) proteins. Essential in endothelial cell migration during vascular development through VEGFA signaling, possibly by regulating RhoA activity. Required for lymphatic vasculature development, possibly by binding to RAS and by activation by EGF and FGF2, but not by PDGF. Regulates invadopodia formation through the PDPK1-AKT1 pathway. Participates in cardiomyogenesis in embryonic stem cells through a AKT1 pathway. Participates in vasculogenesis in embryonic stem cells through PDK1 and protein kinase C pathway. In addition to its lipid kinase activity, it displays a serine-protein kinase activity that results in the autophosphorylation of the p85alpha regulatory subunit as well as phosphorylation of other proteins such as 4EBP1, H-Ras, the IL-3 beta c receptor and possibly others. Plays a role in the positive regulation of phagocytosis and pinocytosis (By similarity).
GenBank BC089038 / GenBank NM_001077495
Especificaciones
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
250 μg | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
Combined 61 to 91% |
129 kDa (p110α) and 83.6 kDa (p85α) | |
Murine | |
N-terminal 6His / Untagged | |
Full-length | |
PI 3-Kinase (p110α/p85α), murine |