missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI3K p110(beta) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20584-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is specific to PIK3CB.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Especificaciones
PI3K p110(beta) | |
Polyclonal | |
Unconjugated | |
PIK3CB | |
p110 BETA, PI3 Kinase p110 beta, PI3 kinase subunit beta, PI3K, PI3K beta, PI3K p110(beta), PI3Kbeta, PI3KCB, PIK3C1, PIK3CB, PtdIns 3 kinase subunit beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5291 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42338 | |
PIK3CB | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |