Learn More
Kern Sohn Plastic Box for Weight Set
Plastic case for individual weight set.
Marca: Kern Sohn 317-020-400
Detalles adicionales : Peso : 0.02000kg
Descripción
- Plastic
- For button/compact weight design
Especificaciones
Case | |
Plastic | |
Class E2 Weights | |
1 g to 2 g weights |