missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 Antibody Cocktail, Invitrogen™
Antibody Cocktail
Marca: Invitrogen 331700
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
33-1700 contains a cocktail of monoclonal mouse PLK1 containing clone PL6, isotype IgG1, kappa and clone PY20, isotype IgG2b, kappa.
PLK1 Cocktail antibody specifically detects PLK1 in Human, Mouse, Non-human Primate, Rat, Xenopus laevis samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Especificaciones
PLK1 | |
Cocktail | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P53350, P70032, Q07832, Q62673 | |
Plk1, plk1.S | |
A His-tagged Plk fusion protein (C-term Plk1) containing amino acids 402-603 of the human Plk1 protein( | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat, X. laevis | |
Antibody | |
IgG1 κ, IgG2b κ |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
PL6/PL2 | |
Unconjugated | |
Plk1 | |
cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg | |
Mouse | |
Protein A | |
RUO | |
18817, 25515, 380481, 5347 | |
-20°C | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido