Learn More
PMP70 Polyclonal Antibody, Alexa Fluor™ 555, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA1650A555
Descripción
PA1650A555 immunizing peptide corresponds to amino acid residues 644-659 from rat PMP70. This sequence is completely conserved in mouse PMP70 and differs from human PMP70 by a single amino acid substitution. The immunizing peptide (Cat. # PEP-038) is available for use in neutralization and control experiments.
Peroxisomes are single membrane organelles which are involved with many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including, phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via beta-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsum"e; disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal beta-oxidation enzymes.
Especificaciones
PMP70 | |
Polyclonal | |
Alexa Fluor 555 | |
ABCD3 | |
68 kDa peroxisomal membrane protein; 70 kDa peroxisomal membrane protein; 70-kDa peroxisomal membrane protein; ABC43; ABCD3; AI313901; ATP binding cassette subfamily D member 3; ATP-binding cassette sub-family D member 3; ATP-binding cassette, subfamily D (ALD), member 3; ATP-binding cassette, sub-family D (ALD), member 3; AU018866; AW146054; CBAS5; dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)); Peroxisomal membrane protein 1; peroxisomal membrane protein 1 (70kD, Zellweger syndrome); peroxisomal membrane protein, 70 kDa; Peroxisomal membrane protein-1 (70kD); PMP68; Pmp70; Pxmp1; ZWS2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19299, 25270, 5825 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P16970, P28288, P55096 | |
ABCD3 | |
Synthetic Peptide: C N(644) Y E F K K I T E D T V E F G S(659) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only