missing translation for 'onlineSavingsMsg'
Learn More

Polimixina B, sulfato, reactivo para cultivo celular, Thermo Scientific Chemicals

Código de producto. p-7076573
Change view
Click to view available options
Cantidad:
1 g
Tamaño de la unidad:
1 g
Código de producto. Cantidad unitSize
15434289 1 g 1 g
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15434289

Marca: Thermo Scientific Alfa Aesar J61763.03

Solicitar formato a granel o personalizado

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Polymixin B sulfate, CAS # 1405-20-5, is a short polypeptide with antibacterial activity against gram-negative bacteria. | CAS: 1405-20-5 | C48H84N16O17S | 1189.36 g/mol

In cell culture, polymyxin B sulfate is used as an antibacterial agent to prevent contamination. Polymyxin B sulfate is recommended for use in cell culture applications at 50 mg/L. It is used as a lipid A binding agent that induces pore formation in cell membranes and as immobilized (substrate bound) agent for removal of endotoxins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Polymixin B sulfate is a mixture of B1 and B2 polypeptides obtained from different strains of Bacillus polymyxa, with antibacterial activity against gram-negative bacteria. It does not exhibit activity against fungi and gram-positive organisms
  • This compound inhibits Ca2+ dependent PKC (protein kinase C)
  • It can bind lipopolysaccharide (LPS) of the outer membrane of gram-negative bacteria and disrupts the cytoplasmic membrane by inducing large pores to allow nucleotide leakage in bacterial walls. This disrupts the permeability of the cytoplasmic membrane

Application

  • Polymixin B sulfate targets BAC-1.2F5 murine macrophages as an LPS (lipopolysaccharide) inhibitor
  • In in vitro research, this compound is used to prevent bacterial contamination of cell cultures
  • In microbiology research, it is used as a positive and negative control during microbicidal concentration (MCC) assay
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1405-20-5
Fórmula molecular C48H84N16O17S
Peso molecular (g/mol) 1189.36
Número MDL MFCD00131991
Clave InChI HNDFYNOVSOOGDU-UHFFFAOYNA-N
PubChem CID 133109994
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O

Especificaciones

Nombre del producto químico o material Polymixin B sulfate
Tipo Polymixin B Sulfate
Forma física Polvo
Envase Frasco de vidrio
Cantidad 1 g
Información de solubilidad Soluble in water
Nombre IUPAC N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoetil)-15-bencil-3-[(1R)-1-hidroxietil]-12-(2-metilpropil)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazaciclotricos-21-il]amino]butan-2-il]amino]-3-hidroxi-1-ox
Formula Weight (peso de la fórmula) 1385.63
compliance-icons
Nombre del producto
  • Polymixin B sulfate, Cell Culture Reagent
Palabra de advertencia
  • Atención
Clasificación
  • Toxicidad aguda Categoría 4
Indicaciones de peligro
  • H302-Nocivo en caso de ingestión.
Consejos de prudencia
  • P301+P312-EN CASO DE INGESTIÓN: Llamar a un CENTRO DE TOXICOLOG A/médico/si la persona se encuentra mal.
Otros peligros
  • MIXTURE LIST-Contém: Polymyxin B sulfate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.