Learn More
BRAND™ 96-well Rack with Tubes
Well-suited for use with robots and other automated dispensing systems. Tubes, racks, and lids can withstand temperatures as low as –80°C. *Cover mat and strips are not autoclavable.
Marca: BRAND™ 781562
Detalles adicionales : Peso : 2.89000kg
Descripción
Tubes and rack are autoclavable at 121°C (2 bar), acc. DIN EN 285 (cover mat/strip, TPE, not autoclavable). Tubes, racks and lids can withstand temperatures as low as -80°C. The seal of the tubes is pressure tested and there is a stable connection between the mounting plate and the racks, making these stackable racks well suited for use with robots and automated systems. Coded tubes (A1-H12) and rack sides are suitable for barcode labels.Ideal for use with robots and other automated dispensing systems.
Especificaciones
With Lid, no tubes |
|
DIN EN 285 |
|
Autoclavable | |
Polypropylene | |
50 Pack |
Tube Rack | |
Yes | |
96 | |
0.65mL Tubes |