Learn More
Thermo Scientific™ POROS™ MabCapture™ A RoboColumn
RoboColumn™ chromatography products are small columns provided in 8-column strips.
356.00€ - 1058.00€
Especificaciones
Tipo de producto | Column |
---|---|
Línea de productos | MABCAPTURE™, POROS™ |
Tipo de columna | Affinity |
Formato | Chromatography Cartridge |
Fase estacionaria | Protein A |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
18436678
|
Thermo Scientific™
A30725 |
200 μL Column |
356.00€
200 microlitros |
Fecha estimada de envío: 23-07-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
18486608
|
Thermo Scientific™
A30726 |
600 μL Column |
1058.00€
600 microlitros |
Fecha estimada de envío: 23-07-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS™ MabCapture™ A affinity resin, which is suitable for monoclonal antibody large-scale downstream bioprocessing.
POROS MabCapture A is a 50-micron affinity resin that combines improvements made in bead and chemistry technology. These advances provide a monoclonal antibody purification resin that not only demonstrates the highest dynamic binding performance at low flow rates, but also one that can maintain superior binding capacity at high linear velocities where other chromatography resins products cannot operate.
Features of the resin:
- Obtain the highest dynamic binding capacity for monoclonal antibodies vs. conventional agarose or silica based Protein A medias
- Maintain high dynamic binding capacity at flow rates over 700 cm/hr
- Design processes with greater flexibility (shorter bed heights, faster flow rates or both) due to the improvements made to the antibody binding efficiency
- Clean and sanitize with standard sodium hydroxide agents utilized for other chromatography products
RoboColumn is a registered trademark of ATOLL GmbH.
Especificaciones
Column | |
Affinity | |
Protein A | |
Store at 2°C to 8°C. |
MABCAPTURE™, POROS™ | |
Chromatography Cartridge | |
Protein | |
Wet Ice |