Learn More
Epredia™ Casetes para biopsia PrintMate™
Código de producto. 15417614 Tienda Epredia Productos
Click to view available options
Color:
Aguamarina
Amarillo
Azul
Beige
Blanco
Canela
Gris
Lavanda
Lila
Naranja
Rosa
Verde
Verde claro
Tamaño de la unidad:
paquete de 1000
paquete de 750
Cantidad:
10 tubos de 75 casetes, 750 por caja en total
Cuatro dispensadores de 250 casetes o 1000 por caja.
Código de producto. 15417614
Marca: Epredia™ A84210053
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Designed for performance and user convenience, the PrintMate cassette range has been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems.  The even, textured printing face provides the optimal surface for high-quality printing every time.  For additional information, the right and left sides are textured for clear handwriting with suitable pens.  The lid catches have been adjusted for ease of use with low force opening and closing, while still giving the security needed while processing.
Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
TRUSTED_SUSTAINABILITY
Especificaciones
Tipo de poro Small Square Pore
Compatible con PrintMate Yes
Color Gris
Tipo de producto Casete para biopsia