missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT3 Antibody (RAB-C372), Janelia Fluor™ 646 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-20143JF646
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PRMT3 Monoclonal antibody specifically detects PRMT3 in Human samples. It is validated for ELISAEspecificaciones
PRMT3 | |
Monoclonal | |
Janelia Fluor 646 | |
EC 2.1.1, EC 2.1.1.-, heterogeneous nuclear ribonucleoprotein methyltransferase-like 3, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HRMT1L3, protein arginine methyltransferase 3, protein arginine N-methyltransferase 3 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT3 protein under non-denaturing conditions. | |
0.1 mL | |
Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
10196 | |
Store at 4°C in the dark. | |
IgG κ |
ELISA | |
RAB-C372 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |