Learn More
PRMT6 Monoclonal Antibody (4G2), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515584
Descripción
MA5-15584 targets PRMT6 in indirect ELISA, WB applications and shows reactivity with Human and Non-human primate samples. The MA5-15584 immunogen is purified recombinant fragment of human PRMT6 expressed in E. Coli. <br>. MA5-15584 detects PRMT6 which has a predicted molecular weight of approximately 42kDa.
Protein arginine N-methyltransferase 6 (PRMT6) PRMT6 demonstrates type I PRMT activity, capable of forming both omega-N monomethylarginine and asymmetric dimethylarginine derivatives on the recombinant glycine- and arginine-rich substrates. cts as a transcriptional coactivator of a number of steroid hormone receptors including ESR1, ESR2, PGR and NR3C1. Promotes fasting-induced transcriptional activation of the gluconeogenic program through methylation of the CRTC2 transcription coactivator. May play a role in innate immunity against HIV-1 in case of infection by methylating and impairing the function of various HIV-1 proteins such as Tat, Rev and Nucleocapsid protein p7.
Especificaciones
PRMT6 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q96LA8 | |
PRMT6 | |
Purified recombinant fragment of human PRMT6 expressed in E. Coli. | |
RUO | |
55170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
4G2 | |
Unconjugated | |
PRMT6 | |
AW124876; BB233495; FLJ10559; FLJ51477; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6; Histone-arginine N-methyltransferase PRMT6; HMT1 hnRNP methyltransferase-like 6; Hrmt1l6; PRMT6; PRMT-6; protein arginine methyltransferase 6; protein arginine N-methyltransferase 6 | |
Mouse | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.