missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Clorhidrato de proflavina
Click to view available options
Cantidad:
5 g
25 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
Vector alfa | PROFLAVINEHYDROCHLORIDE |
Nombre del producto químico o material | Proflavine hydrochloride |
Punto de fusión | 270°C |
Color | Marrón-rojo a naranja |
Pérdida por secado | ≤4 % (105°C, 1 hr) |
Espectro infrarrojo | Conforme |
Intervalo de porcentaje de ensayo | De 13,6 a 15,0 % (CL) (titulación argentométrica) |
Envase | frasco de vidrio |
Cantidad | 25 g |
Información de solubilidad | (1 mg/ml aq. soln.) Clear solution |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'