missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD10 Mouse anti-Human, Clone: OTI3F6, TrueMAB™, Origene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA500781S
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PSMD10 Monoclonal antibody specifically detects PSMD10 in Human samples. It is validated for Flow Cytometry, Western BlotEspecificaciones
PSMD10 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O75832 | |
Mouse | |
Affinity chromatography | |
RUO | |
5716 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
OTI3F6 | |
Unconjugated | |
PSMD10 | |
26S proteasome non-ATPase regulatory subunit 10; 26S proteasome regulatory subunit p28; ankyrin repeat protein; AW554874; dJ889N15.2; Gankyrin; hepatocellular carcinoma-associated protein p28-II; p28; p28(GANK); p28Gank; proteasome (prosome, macropain) 26S subunit, non-ATPase, 10; proteasome 26S subunit, non-ATPase 10; proteasome subunit p28; proteosome (prosome, macropain) 26S subunit, non-ATPase, 10; proteosome subunit p28; PSMD10 | |
Full length human recombinant protein of human PSMD10 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |