Learn More
Julabo™ PURA 4 Water Bath
PURA water baths stand for a straightforward and safe working in the laboratory
Marca:  Julabo™ 9550404
Código de producto. 15646574
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Explore más ofertas especiales
Descripción
Descripción
The high-quality water baths offer a temperature stability of ± 0.15 °C. The working temperature range of +18 °C to 99.9 °C allows a wide range of applications, such as routine laboratory work, temperature applications for samples, incubation, material testing, corrosion testing, temperature control applications of cultures as well as temperature tests of food and beverages and in the dental field.
PURA water baths can be used flexibly and can be easily transported through ergonomic side recessed grips. Non-slip rubber feet ensure a firm footing and more safety in the laboratory.
The units are splash-proof, feature a (membrane) keypad and temperature display (LED) for ease of use. There are no disturbing parts inside the bath so that the entire inner surface can be used to safely temperature-control even small sample containers.
Advantages:
  • Ease of use
  • Bright display
  • Splash-proof protected mains switch
  • Built-in dry running protection
  • No disturbing elements in the bath
  • Platform included as standard
  • Integrated timer
  • Non-slip rubber feet
  • Ergonomic recessed grips
  • Space-saving model for use with one test tube rack
Especificaciones
Especificaciones
Baño de agua
LED Display
35 x 21 x 22 cm
0.8 to 4.5 L
Señal de alarma acústica y óptica
Documentos
One moment while we fetch your results.
Certificados
Ofertas especiales
Ofertas especiales
Título del producto
Julabo™ PURA 4 Water Bath > Capacity (Metric): 0.8 - 4.5 L