Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 10 to 15 in. wide
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
Marca: Keystone Cleanroom Products 6STKRP1036SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
- Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
- Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
- Fast and consistent for equipment preparation area
- Resists tearing and punctures
- Superior microbial barrier
- Manufactured and packaged in a certified ISO 6 cleanroom
- Made in the USA
- Certificates of Conformance sent with shipment
Especificaciones
Autoclave Bag | |
COC | |
Clear, White | |
Esterilización por vapor de equipos farmacéuticos | |
No estéril | |
Sí | |
121 to 125 C | |
36 in | |
10 in |
Autoclavable | |
Safe Seal | |
Single-use | |
Tyvek | |
5.3 mil | |
10 x 36 | |
5,3 mil | |
91.4 cm | |
25.4 cm |