missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVR (CD155) Mouse anti-Human , PE, Clone: SKII.4, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566718
298.03 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Human samples. It is validated for Flow Cytometry
Especificaciones
PVR (CD155) | |
Monoclonal | |
PE | |
poliovirus receptor; HVED; nectin-like protein 5; NECL5; Necl-5; PVR; PVS; TAGE4 | |
SK-N-AS neuroblastoma cell line | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
SKII.4 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
PVR (CD155) Mouse anti-Human , PE, Clone: SKII.4, BD Biosciences™ > 100 Tests; PE
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido