missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCARD Mouse anti-Human, Clone: OTI3E9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA800081S
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PYCARD Monoclonal antibody specifically detects PYCARD in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotEspecificaciones
PYCARD | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9ULZ3 | |
Mouse | |
Affinity chromatography | |
RUO | |
29108 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3E9 | |
Unconjugated | |
PYCARD | |
9130417A21Rik; Apoptosis-associated speck-like protein containing a CARD; Asc; CARD5; caspase recruitment domain-containing protein 5; hASC; mASC; MGC10332; Pycard; PYD and CARD domain containing; PYD and CARD domain-containing protein; Target of methylation-induced silencing 1; TMS; TMS1; TMS-1; TNS1 | |
Full length human recombinant protein of human PYCARD produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |