missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYGL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55429-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is specific to PYGL. It has no cross reaction to PYGM and PYGB.
This gene encodes a homodimeric protein that catalyses the cleavage of alpha-1,4-glucosidic bonds to release glucose-1-phosphate from liver glycogen stores. This protein switches from inactive phosphorylase B to active phosphorylase A by phosphorylation of serine residue 15. Activity of this enzyme is further regulated by multiple allosteric effectors and hormonal controls. Humans have three glycogen phosphorylase genes that encode distinct isozymes that are primarily expressed in liver, brain and muscle, respectively. The liver isozyme serves the glycemic demands of the body in general while the brain and muscle isozymes supply just those tissues. In glycogen storage disease type VI, also known as Hers disease, mutations in liver glycogen phosphorylase inhibit the conversion of glycogen to glucose and results in moderate hypoglycemia, mild ketosis, growth retardation and hepatomegaly. Alternative splicing results in multiple transcript variants encoding different isoforms.Especificaciones
PYGL | |
Polyclonal | |
Unconjugated | |
PYGL | |
GSD6, phosphorylase, glycogen, liver, PYGL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110095, 5836 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06737, Q9ET01 | |
PYGL | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |