missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Lids
For use with SVL™ reaction vessels
Marca: SVL 296-01
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- For use with SVL™ reaction vessels
- Borosilicate glass construction available in three styles: - Plain with knob (no necks) to act as a cover - 296-00 - With two necks (one vertical central neck and one vertical side neck) - With five necks as shown in the table below
Especificaciones
Borosilicate Glass | |
75 mm | |
100 mm |
Reaction Vessel | |
Lid |