missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYREX™ Reusable Glass Tubes with Rubber-Lined Phenolic Caps
Deep-form phenolic caps facilitate handling and sealing after autoclaving
en la oferta
Especificaciones
Color del tapón | Black |
---|---|
Material del cierre | Phenolic |
Tipo de diseño | Threaded |
Material | Borosilicate Glass |
Forma | Round Bottom |
Descripción
Deep-form phenolic caps facilitate handling and sealing after autoclaving
Furnished with deep-form phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of temperature and steam in autoclaving. Caps are provided with a glued-in, white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations. Pencil marks are easily erased.
For phenolic caps, see Cat. No. 9999.
Especificaciones
Black | |
Threaded | |
Round Bottom | |
No | |
Rubber |
Phenolic | |
Borosilicate Glass | |
Non-sterile | |
Marking Spot |