Learn More
Piridina-3,4-dicarboximida, Thermo Scientific Chemicals
Código de producto. 15414387
Click to view available options
Cantidad:
5 g
25 g
100 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 4664-01-1
Número MDL MFCD00013439
Especificaciones
Nombre del producto químico o material Pyridine-3,4-dicarboximide
Punto de fusión 230°C to 232°C
Fórmula molecular C7H4N2O2
Cantidad 25 g
Sinónimo 3,4-pyridinedicarboximide, cinchomeronimide, 3,4-pyridinecarboximide, 1h-pyrrolo 3,4-c pyridine-1,3 2h-dione, 2h-pyrrolo 3,4-c pyridine-1,3-dione, pyrrolo 3,4-c pyridine-1,3-dione, cinchomeronic acid imide, 4-azaphthalimid, pyridine-3,4-dicarboximide, 1h,2h,3h-pyrrolo 3,4-c pyridine-1,3-dione
Clave InChI SJSABZBUTDSWMJ-UHFFFAOYSA-N
SMILES O=C1NC(=O)C2=CN=CC=C12
Nombre IUPAC pirrolo[3,4-c]piridina-1,3-diona
Peso molecular (g/mol) 148.12
PubChem CID 72926
Formula Weight (peso de la fórmula) 148.12