Learn More
R&D Systems™ Recombinant Mouse LRP-6 His Tagged Protein, CF
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Marca:  R&D Systems™ 2960-LR-025
Código de producto. 18158490
  • 390.00€ / 25 microgramos
Explore más ofertas especiales
Descripción
Descripción
The Recombinant Mouse LRP-6 Protein is derived from NS0. The Recombinant Mouse LRP-6 Protein has been validated for the following applications: Binding Activity.
Especificaciones
Especificaciones
Lyophilized from a 0.2μm filtered solution in PBS.
152.6 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
16974
LRP-6
Mouse myeloma cell line,NS0-derived mouse LRP-6 protein Ala20-Pro1368,with a C-terminal 6-His tag
<0.1 EU per 1μg of the protein by the LAL method.
Recombinant
Sugerencias de productos