missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Nectin-4 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 3116-N4-050
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Measured by the ability of the immobilized protein to support the adhesion of NIH-3T3 mouse embryonic fibroblast cells. When 5 x 104 cells/well are added to recombinant mouse Nectin-4 coated plates (10 μg/mL with 100 μL/well), approximately 45%-65% will adhere after 1 hour incubation at 37 °C. Optimal dilutions should be determined by each laboratory for each application.Especificaciones
NP_082169 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 40-50 kDa, reducing conditions; M.W. (theoretical): 35.6 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 500 μg/mL in PBS. | |
Nectin-4/PVRL4 |
Bioactivity | |
81607 | |
Nectin-4 | |
Mouse myeloma cell line, NS0-derived mouse Nectin-4 protein Tyr28-Ile349, with a C-terminal 6-His tag | |
Monomer | |
Nectin4 | |
Recombinant | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido