Learn More
R&D Systems™ Recombinant Human MMP-8 Western Blot Standard Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot
Marca: R&D Systems™ WBC017
117.49 EUR hasta el 2024-12-16
Use el código promocional "21630" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Human MMP-8 Western Blot Standard Protein has been validated for the following applications: Western Blot.Especificaciones
Western Blot | |
MMP-8 Western Blot Standard | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Unconjugated | |
MMP-8 |
4317 | |
1 Vial | |
MMP8 | |
Recombinant |
Corrección del contenido de un producto