Learn More
RAB17 Monoclonal Antibody (OTI5E2), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA525117
Descripción
The Rab subfamily of small GTPases plays an important role in the regulation of membrane trafficking. RAB17 is an epithelial cell-specific GTPase.
Especificaciones
RAB17 | |
Monoclonal | |
0.63 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H0T7 | |
Rab17 | |
Full length human recombinant protein of RAB17 produced in HEK293T cell | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI5E2 | |
Unconjugated | |
Rab17 | |
AW413472; Rab17; RAB17, member RAS oncogene family; ras-related protein Rab-17; RGD1562478 | |
Mouse | |
Affinity Chromatography | |
RUO | |
64284 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.