missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Horse IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™

Marca:  Abnova PAB10662.2mg

Detalles adicionales : Peso : 0.00010kg

Código de producto. 16242970

  • 396.00€ / 2 miligramos
Fecha estimada de envío: 28-08-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

Rabbit anti-Horse IgG whole molecule
Polyclonal
Rabbit anti-horse IgG recognizes horse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% gentamicLyophilized from sulfate)
Horse IgG whole molecule
2 mg
Equine
Lyophilized
Dot Blot, ELISA, Immunohistochemistry, Western Blot
HRP
The optimal working dilution should be determined by the end user.
Rabbit
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only