Learn More
RAF1(phospho S259) & RAF1 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Marca:  Abnova DP0129.1Set
Código de producto. 16131604
  • 713.00€ / 1 set
Explore más ofertas especiales
Descripción
Descripción
This protein phosphorylation antibody pair set comes with two antibodies, one against the RAF1 protein, and the other against the specific S259 phosphorylated site of RAF1 for use in in situ Proximity Ligation Assay.
Especificaciones
Especificaciones
RAF1
Unconjugated
CRAF/NS5/Raf-1/c-Raf
1 Set
Primary
Antibody pair set content: 1. Phospho-RAF1 S259 rabbit polyclonal antibody (20 ul) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. RAF1 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
In situ PLA
v-raf-1 murine leukemia viral oncogene homolog 1
RAF1
RUO
5894
Human
Ab Pair
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
RAF1(phospho S259) & RAF1 Protein Phosphorylation Antibody Pair, Abnova™ > 1 Set; Unconjugated