Learn More
CD4 Rat anti-Mouse, Brilliant Violet 510, Clone: RM4-4, BD Optibuild™
Rat Monoclonal Antibody
Marca: BD Biosciences 740105
Descripción
The RM4-4 monoclonal antibody specifically binds to CD4 (L3T4) differentiation antigen expressed on most thymocytes, a subpopulation of mature T lymphocytes (ie, MHC class II-restricted T cells, including most T helper cells), and a subset of NK-T cells of all mouse strains tested. CD4 has also been detected at low density on pluripotent hematopoietic stem cells, bone marrow myeloid and B-lymphocyte precursors, intrathymic lymphoid precursors, and a subset of splenic dendritic cells. CD4 is expressed on the plasma membrane of mouse egg cells and is involved in adhesion of the egg to MHC class II-bearing sperm. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. Purified RM4-4 mAb does not block binding of the CD4-specific monoclonal antibodies, GK1.5, H129.19, or RM4-5, to T cells.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.
Especificaciones
| CD4 | |
| Monoclonal | |
| 0.2mg/mL | |
| Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| RM4-4 | |
| Brilliant Violet 510 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| BALB/c mouse thymocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only