missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat Calcium Sensing Receptor (aa 12-27) Synthetic Peptide

Synthetic Peptide

Marca:  Invitrogen™ PEP009

Detalles adicionales : CAS : 69-74-9 Peso : 0.01000kg

Código de producto. 11528881

  • 167.00€ / 50 microgramos
Fecha estimada de envío: 12-08-2024
para ver el stock

Descripción

Descripción

Rat Calcium-Sensing Receptor (residues 12-27) Synthetic Peptide, PEP-009, from Invitrogen.

PEP-009 is a 16 amino acid (amino acids 12-27) synthetic peptide whose sequence is from the amino-terminal end of the rat calcium-sensing receptor protein. The sequence of this peptide is (amino to carboxy terminus): A-L-A-W-H-S-S-A-Y-G-P-D-Q-R-A-QThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and the calcium-sensing receptor, catalog # PA1-934A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
Especificaciones

Especificaciones

24247
purified
Rat Calcium Sensing Receptor (aa 12-27)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Rat
P48442
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
Casr
No preservative
50 μg
A - L - A - W - H - S - S - A - Y - G - P - D - Q - R - A - Q
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales