Learn More
ACROBiosystems Rat CD47 Protein, Fc Tag
350.80€ - 2419.31€
Especificaciones
Peso molecular | 40.4 kDa |
---|---|
Fuente | Rat CD47, Fc Tag (CD7-R5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # P97829). |
Inmunógeno | Gln 19 - Lys 140 |
Requisitos de almacenamiento | -20°C |
Alias de gen | CD47,MER6,IAP,OA3 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15896287
|
ACROBiosystems
CD7-R5256-100UG |
100 μg |
350.80€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15806297
|
ACROBiosystems
CD7-R5256-1MG |
1 mg |
2419.31€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Leukocyte surface antigen CD47 is also known as Antigenic surface determinant protein OA3, Integrin-associated protein (IAP) and Protein MER6. CD47 contains 1 Ig-like V-type (immunoglobulin-like) domain. CD47 is very broadly distributed on normal adult tissues. CD47 has a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins and plays an important role in memory formation and synaptic plasticity in the hippocampus by similarity. CD47 is the receptor for SIRPA, binding to which prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells. CD47 Interaction with SIRPG mediates cell-cell adhesion, enhances superantigen-dependent T-cell-mediated proliferation and costimulates T-cell activation.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. As a result of glycosylation, the protein migrates as 55-67 kDa under reducing (R) condition, and 100-130 kDa under non-reducing (NR) condition (SDS-PEspecificaciones
40.4 kDa | |
Gln 19 - Lys 140 | |
CD47,MER6,IAP,OA3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Rat CD47, Fc Tag (CD7-R5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # P97829). | |
-20°C | |
CD47 | |
Rat | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |