missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat IL-2 Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
50 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
50 microgramos
500 microgramos
Description
Recombinant Rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
Specifications
| Número de acceso | P17108 |
| Para utilizar con (aplicación) | ELISA, Functional Assay, Western Blot |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 116562 |
| Peso molecular | 15.3 kDa |
| Nombre | Rat IL-2 |
| Cantidad | 50 μg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction