missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RbAp48 Monoclonal Antibody (13D10), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA123275
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: HeLa nuclear extract, SH-SY-5Y, SH-SY-5Y nuclear extract.
RbAp48-11G10 recognizes full length RbAp48, a 48 kDa nuclear protein first isolated by its ability to interact with the carboxyl-terminus of the retinoblastoma protein (Rb). It is a member of the WD (Trp-Asp) repeat family of proteins. RbAp48 is one of the three subunits of CAF-1, can bind to histone H4 and is a subunit of human histone deacetylase HD1.Especificaciones
RbAp48 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q09028, Q60972 | |
RBBP4 | |
The complete coding region (amino acids 1-425) of RbAp48 expressed in E. coli. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
13D10 | |
Unconjugated | |
RBBP4 | |
CAF-1 p48 subunit; CAF-1 subunit C; CAF-I 48 kDa subunit; CAF-I p48; chromatin assembly factor 1 subunit C; Chromatin assembly factor I p48 subunit; chromatin assembly factor/CAF-1 p48 subunit; histone-binding protein RBBP4; lin-53; mRbAp48; MSI1 protein homolog; nucleosome-remodeling factor subunit RBAP48; NURF55; RB binding protein 4, chromatin remodeling factor; RBAP48; RBBP4; RBBP-4; retinoblastoma binding protein 4; retinoblastoma-binding protein 4; Retinoblastoma-binding protein p48 | |
Mouse | |
Protein G | |
RUO | |
19646, 313048, 5928 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |