missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBPJ Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA522226
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: 293T, A431, HeLa, Molt-4, Raji. Predicted reactivity: Mouse (97%), Rat (98%), Xenopus laevis (93%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.
Especificaciones
| RBPJ | |
| Polyclonal | |
| Unconjugated | |
| RBPJ | |
| AI843960; AOS3; CBF1; CBF-1; csl; H-2K binding factor-2; Igkjrb; Igkjrb1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; rbp-jkappa; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; recombining binding protein suppressor of hairless-like protein; renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog; Unknown (protein for MGC:128782) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3516 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.75 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q06330 | |
| RBPJ | |
| Recombinant protein encompassing a sequence within the center region of human RBPJK. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido