Learn More
BUCHI Recirculating Chillers
BUCHI Recirculating Chillers are easy to use and designed for use with Rotavapors, SyncorePlus, Multivapor, Kjeldahl systems, Extraction systems, and other suppliers' similar products.
20735.00€
Código de producto | Marca | Descripción | Para utilizar con (equipo) | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Descripción | Para utilizar con (equipo) | Precio | Cantidad y disponibilidad | ||||
16824590
|
BUCHI
11F32501 |
Coooling Capacity: 2500w at 15°C | Rotavapor R-220 Pro Industrial Evaporator |
20735.00€
cada uno |
Fecha estimada de envío: 05-12-2024
Inicie sesión para ver el stock |
||||
Descripción
- Efficient distillation due to complete integration into the BUCHI rotary evaporation system
- Temperature lock and temperature control in conjunction with the Interface I-300 / I-300 Pro.
- Digital Display: Set and actual temperature
- Saves time thanks to instant start with automatic and dynamic pressure adjustment without waiting until chiller reaches its set temperature
- ECO-mode: Saves energy and reduces heat emission by switching into a standby mode when not in use
- No water consumption
- Maximizes distillation capacity while reducing solvent emissions due to intelligent integration of all process parameters using the Interface I-300 / I-300 Pro
- Seamless plug and play integration into:
- Fully automated BUCHI Rotavapor R-300, R-220 Pro systems including all process parameters
- BUCHI parallel evaporators, extraction and Kjeldahl solutions