Learn More
BUCHI Recirculating Chillers
BUCHI Recirculating Chillers are easy to use and designed for use with Rotavapors, SyncorePlus, Multivapor, Kjeldahl systems, Extraction systems, and other suppliers' similar products.
20735.00€
Productos 1
Código de producto Marca Descripción Para utilizar con (equipo) Precio Cantidad y disponibilidad  
Código de producto Marca Descripción Para utilizar con (equipo) Precio Cantidad y disponibilidad  
16824590
Ver documentos Detalles de la promoción
BUCHI
11F32501
Coooling Capacity: 2500w at 15°C Rotavapor R-220 Pro Industrial Evaporator
20735.00€
cada uno
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 05-12-2024
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • Efficient distillation due to complete integration into the BUCHI rotary evaporation system
  • Temperature lock and temperature control in conjunction with the Interface I-300 / I-300 Pro.
  • Digital Display: Set and actual temperature
  • Saves time thanks to instant start with automatic and dynamic pressure adjustment without waiting until chiller reaches its set temperature
  • ECO-mode: Saves energy and reduces heat emission by switching into a standby mode when not in use
  • No water consumption
  • Maximizes distillation capacity while reducing solvent emissions due to intelligent integration of all process parameters using the Interface I-300 / I-300 Pro
  • Seamless plug and play integration into:
    • Fully automated BUCHI Rotavapor R-300, R-220 Pro systems including all process parameters
    • BUCHI parallel evaporators, extraction and Kjeldahl solutions