missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant B. abortus vjbR Protein
A DNA sequence encoding the Brucella abortus (strain 2308) vjbR, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7642-ye-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
WP_002966460.1 | |
vjbR Protein | |
Research Use Only | |
vjbR | |
Recombinant Protein | |
Yeast | |
MSLDLVHFPN YKKTFFGSSF QSDTLALLTR IRDEIGCRYV THTYRGRVGD CTKVNSADLT VLMTLPATWV ARYSSKNYFA IDPVFQEDAP YYRNDTSAIA RDLKEDADIC PAVAELLHDA EKHGLGNLFI AVSARNPKGV AGCTVFTFEV EDEDRTQFLA RMRPRLLSLA GIIHGTVCGC KDANSVASLL TPREVDCLRW AANGKTDGEI AEILSIARWT VVTYLQNAKI KLNCSNRTSA VATALSLGII DMPEVQHLV | |
Greater than 90% as determined by SDS-PAGE. |
29595784 | |
1 mg | |
Not determined. | |
Full Length | |
B. abortus | |
His | |
Tris-based buffer, 50% glycerol |