Learn More
R&D Systems™ Recombinant Canine Erythropoietin/EPO Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 3816-CE-010
285.09 EUR hasta el 2024-12-16
Use el código promocional "21630" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Canine Erythropoietin Protein is derived from NS0. The Recombinant Canine Erythropoietin Protein has been validated for the following applications: Bioactivity.Especificaciones
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
Erythropoietin/EPO | |
Mouse myeloma cell line, NS0-derived canine Erythropoietin/EPO protein Ala41-Arg206 | |
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
Recombinant |
18.4 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |