missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog CMA1 / Chymase 1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) CMA1 / Chymase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5843-ye-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001013442.1 | |
CMA1 / Chymase 1 Protein | |
Research Use Only | |
CMA1 | |
Recombinant Protein | |
Yeast | |
IIGGTESKPH SRPYMAHLEI LTLRNHLASC GGFLIRRNFV LTAAHCAGRF IMVTLGAHNI QKKEDTWQKL EVIKQFPHPK YDDLTLRHDI MLLKLKEKAN LTLAVGTLPL SPQFNFVPPG RMCRVAGWGK RQVNGSGSDT LQEVKLRLMD PQACRHYMAF DHNLQLCVGN PRKTKSAFKG DSGGPLLCAG VAQGIVSYGQ NDAKPPAVFT RISHYRPWIN KVLKQNKA | |
Greater than 90% as determined by SDS-PAGE. |
490628 | |
100 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |