missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glutaminase 1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Glutaminase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6951-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_415018.1 | |
Glutaminase 1 Protein | |
Research Use Only | |
glsA1 | |
Recombinant Protein | |
E. coli | |
MLDANKLQQA VDQAYTQFHS LNGGQNADYI PFLANVPGQL AAVAIVTCDG NVYSAGDSDY RFALESISKV CTLALALEDV GPQAVQDKIG ADPTGLPFNS VIALELHGGK PLSPLVNAGA IATTSLINAE NVEQRWQRIL HIQQQLAGEQ VALSDEVNQS EQTTNFHNRA IAWLLYSAGY LYCDAMEACD VYTRQCSTLL NTIELATLGA TLAAGGVNPL THKRVLQADN VPYILAEMMM EGLYGRSGDW AYRVGLPGKS GVGGGILAVV PGVMGIAAFS PPLDEDGNSV RGQKMVASVA KQLGYNVFKG | |
Greater than 90% as determined by SDS-PAGE. |
946187 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His-GST | |
Tris-based buffer, 50% glycerol |