missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Hygromycin-B 4-O-kinase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Hygromycin-B 4-O-kinase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7397-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
WP_000742814.1 | |
Hygromycin-B 4-O-kinase Protein | |
Research Use Only | |
hph | |
Recombinant Protein | |
E. coli | |
MKKPELTATS VEKFLIEKFD SVSDLMQLSE GEESRAFSFD VGGRGYVLRV NSCADGFYKD RYVYRHFASA ALPIPEVLDI GEFSESLTYC ISRRAQGVTL QDLPETELPA VLQPVAEAMD AIAAADLSQT SGFGPFGPQG IGQYTTWRDF ICAIADPHVY HWQTVMDDTV SASVAQALDE LMLWAEDCPE VRHLVHADFG SNNVLTDNGR ITAVIDWSEA MFGDSQYEVA NIFFWRPWLA CMEQQTRYFE RRHPELAGSP RLRAYMLRIG LDQLYQSLVD GNFDDAAWAQ GRCDAIVRSG AGTVGRTQIA RRSAAVWTDG CVEVLADSGN RRPSTRPRAK E | |
Greater than 90% as determined by SDS-PAGE. |
13905269 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |