missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Latent membrane protein 1
A DNA sequence encoding the Epstein-Barr virus (strain Raji) (HHV-4) (Human herpesvirus 4) Latent membrane protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7024-ye-50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Latent membrane protein 1 | |
Research Use Only | |
LMP1 | |
Recombinant Protein | |
Yeast | |
YYHGQRHSDE HHHDDSLPHP QQATDDSSNQ SDSNSNEGRH LLLVSGAGDG PPLCSQNLGA PGGGPNNGPQ DPDNTDDNGP QDPDNTDDNG PHDPLPQDPD NTDDNGPQDP DNTDDNGPHD PLPHNPSDSA GNDGGPPQLT EEVENKGGDQ GPPLMTDGGG GHSHDSGHDG IDPHLPTLLL GTSGSGGDDD DPHGPVQLSY YD | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
Epstein-Barr | |
His | |
Tris-based buffer, 50% glycerol |