missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBeAg Protein
A cDNA sequence encoding the HBeAg was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12120-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis B HBeAg Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SKLCLGWLWG MDIDPYKEFG ATVELLSFLP SDFFPSVRDL LDTASALYRE ALESPEHCSP HHTALRQAIL CWGELMTLAT WVGNNLEDPA SRDLVVNYVN TNVGLKIRQL LWFHISCLTF GRETVLEYLV SFGVWIRTPP AYRPPNAPIL STLPETTVVR RRDRGRSPRR RTPSPRRRRS PSPRRRRSQS RESQC | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
His | |
Phosphate buffered saline with 50mM arginine. |