missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis B HBeAg Protein

A cDNA sequence encoding the HBeAg was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12120-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15999428

  • 4090.00€ / 1 miligramo
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

Hepatitis B HBeAg Protein
Research Use Only
Recombinant Protein
E. coli
SKLCLGWLWG MDIDPYKEFG ATVELLSFLP SDFFPSVRDL LDTASALYRE ALESPEHCSP HHTALRQAIL CWGELMTLAT WVGNNLEDPA SRDLVVNYVN TNVGLKIRQL LWFHISCLTF GRETVLEYLV SFGVWIRTPP AYRPPNAPIL STLPETTVVR RRDRGRSPRR RTPSPRRRRS PSPRRRRSQS RESQC
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis B
His
Phosphate buffered saline with 50mM arginine.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales