missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse Serum amyloid A Protein
A DNA sequence encoding the Equus caballus (Horse) Serum amyloid A protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6647-ye-500ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Serum amyloid A Protein | |
Research Use Only | |
SAA1 | |
Recombinant Protein | |
Yeast | |
LLSFLGEAAR GTWDMIRAYN DMREANYIGA DKYFHARGNY DAAKRGPGGA WAAKVISDAR ENFQRFTDRF SFGGSGRGAE DSRADQAANE WGRSGKDPNH FRPHGLPDKY | |
Greater than 80% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Horse | |
His | |
Tris-based buffer, 50% glycerol |