missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 2-5A-dependent ribonuclease Protein
A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C virus genotype 1a (isolate 1) polyprotein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6997-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_066956.1 | |
2-5A-dependent ribonuclease Protein | |
Research Use Only | |
RNASEL | |
Recombinant Protein | |
E. coli | |
HPPAEDWKPQ SSHWGAALKD LHRIYRPMIG KLKFFIDEKY KIADTSEGGI YLGFYEKQEV AVKTFCEGSP RAQREVSCLQ SSRENSHLVT FYGSESHRGH LFVCVTLCEQ TLEACLDVHR GEDVENEEDE FARNVLSSIF KAVQELHLSC GYTHQDLQPQ NILIDSKKAA HLADFDKSIK WAGDPQEVKR DLEDLGRLVL YVVKKGSISF EDLKAQSNEE VVQLSPDEET KDLIHRLFHP GEHVRDCLSD LLGHPFFWTW ESRYRTLRNV GNESDIKTRK SESEILRLLQ PGPSEHSKSF DKWTTKINEC VMKKMNKFYE KRGNFYQNTV GDLLKFIRNL GEHIDEEKHK KMKLKIGDPS LYFQKTFPDL VIYVYTKLQN TEYRKHFPQT HSPNKPQCDG AGGASGLASP GC | |
Greater than 90% as determined by SDS-PAGE. |
6041 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |