missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human a-N-acetylgalactosaminidase Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 6717-GH-020
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Measured by its ability to cleave alpha-N-acetylgalactosaminyl from 4-Nitrophenyl N-acetyl-alpha-D-galactosaminide. The specific activity is >1800 pmol/min/μg, as measured under the described condition.Especificaciones
P17050 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl, Brij and Glycerol. | |
M.W. (Observed): 50-60 kDa, reducing conditions; M.W. (theoretical): 46 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
NAGA | |
Recombinant | |
alpha-N-acetylgalactosaminidase/NAGA |
Bioactivity | |
4668 | |
alpha-N-acetylgalactosaminidase/NAGA | |
Chinese Hamster Ovary cell line, CHO-derived human alpha-N-acetylgalactosaminidase/NAGA protein Leu18-Gln411, with a C-terminal 6-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.) |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido