missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AGO2 / Argonaute 2 / EIF2C2 Protein
A DNA sequence encoding the Homo sapiens (Human) AGO2 / Argonaute 2 / EIF2C2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8243-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001158095.1 | |
AGO2 / Argonaute 2 / EIF2C2 Protein | |
Research Use Only | |
AGO2 | |
Recombinant Protein | |
E. coli | |
LVVVILPGKT PVYAEVKRVG DTVLGMATQC VQMKNVQRTT PQTLSNLCLK INVKLGGVNN ILLPQGRPPV FQQPVIFLGA DVTHPPAGDG KKPSIAAVVG SMDAHPNRYC ATVRVQQHRQ EIIQDLAAMV RELLIQFYKS TRFKPTRIIF YRDGVSEGQF QQVLHHELLA IREACIKLEK DYQPGITFIV VQKRHHTRLF CTDKNERVGK SGNIPAGTTV DTKITHPTEF DFYLCSHAGI QGTSRPSHYH VLWDDNRFSS DELQILTYQL CHTYVRCTRS VSIPAPAYYA HLVAFRARYH LV | |
Greater than 80% as determined by SDS-PAGE. |
27161 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |