missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Carnosine Dipeptidase 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marca: R&D Systems™ 2489-ZN-010
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Measured by its ability to cleave carnosine ( beta-Ala-L-His) in a two-step assay. The specific activity is >250 pmol/min/μg, as measured under the described conditions.Especificaciones
Q96KN2 | |
Supplied as a 0.2 μm filtered solution in Tris and NaCl. | |
M.W. (Observed): 65 kDa, reducing conditions; M.W. (theoretical): 55 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
CNDP1 | |
Recombinant | |
Carnosine Dipeptidase 1/CNDP1 |
Enzyme Activity | |
84735 | |
Carnosine Dipeptidase 1/CNDP1 | |
Mouse myeloma cell line, NS0-derived human Carnosine Dipeptidase 1/CNDP1 protein Pro28-His507, with a C-terminal 10-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido