missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Casein Kinase 2 alpha Protein
Human Recombinant Protein
Marca: R&D Systems™ 7957-CK-020
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Human Casein Kinase 2 alpha Protein is derived from E. coli.Especificaciones
P68400-1 | |
1457 | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Casein Kinase 2 alpha | |
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Supplied as a 0.2 μm filtered solution in MES, NaCl. | |
M.W. (Observed): 14-16 kDa, reducing conditions; M.W. (theoretical): 16 kDa | |
E. coli-derived human Casein Kinase 2 alpha protein Asp253-Gln391, with an N-terminal Met and C-terminal 6-His tag | |
RUO | |
Unconjugated | |
Human | |
Casein Kinase 2 alpha |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido