missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD72 Protein
A cDNA sequence encoding the CD72 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11345-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
971 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAEAITYADL RFVKAPLKKS ISSRLGQDPG ADDDGEITYE NVQVPAVLGV PSSLASSVLG DKAAVKSEQP TASWRAVTSP AVGRILPCRT TCLRYLLLGL LLTCLLLGVT AICLGVRYLQ VSQQLQQTNR VLEVTNSSLR QQLRLKITQL GQSAEDLQGS RRELAQSQEA LQVEQRAHQA AEGQLQACQA DRQKTKETLQ SEEQQRRALE QKLSNMENRL KPFFTCGSAD TCCPSGWIMH QKSCFYISLT SKNWQESQKQ CETLSSKLAT FSEIYPQSHS YYFLNSLLPN GGSGNSYWTG LSSNKDWKLT DDTQRTRTYA QSSKCNKVHK TWSWWTLESE SCRSSLPYIC EMTAFRFPD |
CD72 Protein | |
Research Use Only | |
CD72 | |
Human | |
Untagged | |
CD72 at 100microg/ml in 50mM Tris-HCl, pH 7.5, 10mM L-glutathione (reduced). |